accessibility menu, dialog, popup

UserName

CAS RN 93479-97-1

H3CH3CNOHNSOONHONHCH3O

CAS RN 93479-97-1

IUPAC Name: 3-ethyl-4-methyl-N-{2-[4-({[(4-methylcyclohexyl)carbamoyl]amino}sulfonyl)phenyl]ethyl}-2-oxo-2,5-dihydro-1H-pyrrole-1-carboxamide
Synonyms: glimepiride glimepiride (product) aventis brand of glimepiride glimepiride preparation roname pms-glimepiride glimepiride (substance) 格列美脲 lacer brand of glimepiride aventis pharma brand of glimepiride
Molecular Weight (g/mol): 490.62
Molecular Formula: C24H34N4O5S
InChi Key: WIGIZIANZCJQQY-UHFFFAOYSA-N
SMILES: CCC1=C(C)CN(C(=O)NCCC2=CC=C(C=C2)S(=O)(=O)NC(=O)NC2CCC(C)CC2)C1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Certifications/Compliance 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)