CAS RN 934570-60-2
CAS RN 934570-60-2
IUPAC Name:
tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
Synonyms:
tert-butyl 2-chloro-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol):
219.68
Molecular Formula:
C8H10ClNO2S
InChi Key:
SQZXVWDARSIXTM-UHFFFAOYSA-N
SMILES:
CC(C)(C)OC(=O)C1=CN=C(Cl)S1
1
–
1
of
1
results
1
tert-Butyl 2-chloro-1,3-thiazole-5-carboxylate, 95%, Thermo Scientific™
CAS: 934570-60-2 Molecular Formula: C8H10ClNO2S Molecular Weight (g/mol): 219.683 MDL Number: MFCD09879991 InChI Key: SQZXVWDARSIXTM-UHFFFAOYSA-N Synonym: tert-butyl 2-chlorothiazole-5-carboxylate,t-butyl-2-chloro-1,3-thiazole-5-carboxylate PubChem CID: 24229786 IUPAC Name: tert-butyl 2-chloro-1,3-thiazole-5-carboxylate SMILES: CC(C)(C)OC(=O)C1=CN=C(S1)Cl