accessibility menu, dialog, popup

UserName

CAS RN 933986-97-1

BOOH3CCH3H3CH3CNN

CAS RN 933986-97-1

IUPAC Name: 2-(pyrrolidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Synonyms: 2-(pyrrolidin-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Molecular Weight (g/mol): 274.17
Molecular Formula: C15H23BN2O2
InChi Key: HGBBKKWVSQBIMM-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CN=C(C=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results