accessibility menu, dialog, popup

UserName

CAS RN 933674-93-2

CH3OFFFNH2

CAS RN 933674-93-2

IUPAC Name: 2-methyl-5-(trifluoromethoxy)aniline
Synonyms: 2-methyl-5-(trifluoromethoxy)aniline
Molecular Weight (g/mol): 191.15
Molecular Formula: C8H8F3NO
InChi Key: WSUXTASOGISSKI-UHFFFAOYSA-N
SMILES: CC1=C(N)C=C(OC(F)(F)F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results