accessibility menu, dialog, popup

UserName

OFFFOOH

CAS RN 93249-62-8

IUPAC Name: 2-hydroxy-5-(trifluoromethoxy)benzaldehyde
Synonyms: 2-hydroxy-5-(trifluoromethoxy)benzaldehyde
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: WQUZBERVMUEJTD-UHFFFAOYSA-N
SMILES: OC1=C(C=O)C=C(OC(F)(F)F)C=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)