accessibility menu, dialog, popup

UserName

CAS RN 932-01-4

CH3H3COH

CAS RN 932-01-4

IUPAC Name: 4,4-dimethylcyclohexan-1-ol
Synonyms: 4,4-dimethylcyclohexan-1-ol
Molecular Weight (g/mol): 128.22
Molecular Formula: C8H16O
InChi Key: VUQOIZPFYIVUKD-UHFFFAOYSA-N
SMILES: CC1(C)CCC(O)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results