accessibility menu, dialog, popup

UserName

CAS RN 930-68-7

O

CAS RN 930-68-7

IUPAC Name: cyclohex-2-en-1-one
Synonyms: cyclohexenone 18O-labeled 2-cyclohexen-1-one 18o-labeled 2-cyclohexen-1-one 2-cyclohexen-1-one, ion(1-) ion(1-) 2-cyclohexen-1-one
Molecular Weight (g/mol): 96.13
Molecular Formula: C6H8O
InChi Key: FWFSEYBSWVRWGL-UHFFFAOYSA-N
SMILES: O=C1CCCC=C1
Quantity 
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)
  • (3)
  • (4)
  • (4)