accessibility menu, dialog, popup

UserName

CAS RN 930-51-8

CH

CAS RN 930-51-8

IUPAC Name: ethynylcyclopentane
Synonyms: ethynylcyclopentane
Molecular Weight (g/mol): 94.16
Molecular Formula: C7H10
InChi Key: TXVJSWLZYQMWPC-UHFFFAOYSA-N
SMILES: C#CC1CCCC1
Boiling Point 
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (5)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)