accessibility menu, dialog, popup

UserName

CAS RN 930-37-0

CH3OO

CAS RN 930-37-0

IUPAC Name: 2-(methoxymethyl)oxirane
Synonyms: oxirane, (methoxymethyl)-
Molecular Weight (g/mol): 88.11
Molecular Formula: C4H8O2
InChi Key: LKMJVFRMDSNFRT-UHFFFAOYNA-N
SMILES: COCC1CO1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results