accessibility menu, dialog, popup

UserName

CAS RN 93-54-9

CH3OH

CAS RN 93-54-9

IUPAC Name: 1-phenylpropan-1-ol
Synonyms: 1-phenyl-1-propanol phenychol phenycholon livonal ejibil fepar felitrope felicur phenylchol �-ethylbenzyl alcohol
Molecular Weight (g/mol): 136.19
Molecular Formula: C9H12O
InChi Key: DYUQAZSOFZSPHD-UHFFFAOYNA-N
SMILES: CCC(O)C1=CC=CC=C1
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)