CAS RN 93-20-9
CAS RN 93-20-9
IUPAC Name:
2-(naphthalen-2-yloxy)ethan-1-ol
Synonyms:
2-(2-naphthyloxy)ethanol
Molecular Weight (g/mol):
188.23
Molecular Formula:
C12H12O2
InChi Key:
BQPBZDSDFCDSAO-UHFFFAOYSA-N
SMILES:
OCCOC1=CC=C2C=CC=CC2=C1
1
–
1
of
1
results
1
2-(2-Naphthoxy)ethanol, 98+%
CAS: 93-20-9 Molecular Formula: C12H12O2 Molecular Weight (g/mol): 188.226 MDL Number: MFCD00016809 InChI Key: BQPBZDSDFCDSAO-UHFFFAOYSA-N Synonym: 2-2-naphthyloxy ethanol,2-2-naphthoxy ethanol,2-naphthalen-2-yloxy ethanol,anavenol,2-2-hydroxyethoxy naphthalene,ethanol, 2-2-naphthalenyloxy,2-2-naphthalenyloxy ethanol,unii-7m7cut7ccu,ethylene glycol mono-2-naphthyl ether,.beta.-naphthoxyethanol PubChem CID: 7131 IUPAC Name: 2-naphthalen-2-yloxyethanol SMILES: C1=CC=C2C=C(C=CC2=C1)OCCO