accessibility menu, dialog, popup

UserName

CAS RN 929-73-7

ClH3CNH3

CAS RN 929-73-7

IUPAC Name: dodecan-1-aminium chloride
Synonyms: 1-dodecanamine, hydrochloride
Molecular Weight (g/mol): 221.81
Molecular Formula: C12H28ClN
InChi Key: TWFQJFPTTMIETC-UHFFFAOYSA-N
SMILES: [Cl-].CCCCCCCCCCCC[NH3+]
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)