accessibility menu, dialog, popup

UserName

CAS RN 929-17-9

H2NOHO

CAS RN 929-17-9

IUPAC Name: 7-aminoheptanoic acid
Synonyms: 7-aminoheptanoic acid
Molecular Weight (g/mol): 145.2
Molecular Formula: C7H15NO2
InChi Key: XDOLZJYETYVRKV-UHFFFAOYSA-N
SMILES: NCCCCCCC(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)