CAS RN 92855-65-7
IUPAC Name:
7-(benzyloxy)-1H-indole-3-carbaldehyde
Synonyms:
7-(benzyloxy)-1H-indole-3-carbaldehyde
Molecular Weight (g/mol):
251.29
Molecular Formula:
C16H13NO2
InChi Key:
RNARWQXZOKWYHY-UHFFFAOYSA-N
SMILES:
O=CC1=CNC2=C(OCC3=CC=CC=C3)C=CC=C12
1
–
1
of
1
results
1
Alfa Aesar™ 7-Benzyloxyindole-3-carboxaldehyde, 97%
CAS: 92855-65-7 Molecular Formula: C16H13NO2 Molecular Weight (g/mol): 251.285 MDL Number: MFCD00055979 InChI Key: RNARWQXZOKWYHY-UHFFFAOYSA-N Synonym: 7-benzyloxyindole-3-carbaldehyde,7-benzyloxy-3-formylindole,7-benzyloxy-1h-indole-3-carbaldehyde,7-benzyloxyindole-3-carboxaldehyde,pubchem14131,7-phenylmethoxy indole-3-carbaldehyde,7-benzyloxy-1h-indole-3-carboxaldehyde,7-benzyloxindole-3-carboxaldehyde,1h-indole-3-carboxaldehyde, 7-phenylmethoxy PubChem CID: 260799 IUPAC Name: 7-phenylmethoxy-1H-indole-3-carbaldehyde SMILES: C1=CC=C(C=C1)COC2=CC=CC3=C2NC=C3C=O