accessibility menu, dialog, popup

UserName

CAS RN 928-98-3

HSSH

CAS RN 928-98-3

IUPAC Name: pentane-1,5-dithiol
Synonyms: pentane-1,5-dithiol
Molecular Weight (g/mol): 136.27
Molecular Formula: C5H12S2
InChi Key: KMTUBAIXCBHPIZ-UHFFFAOYSA-N
SMILES: SCCCCCS
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)