accessibility menu, dialog, popup

UserName

CAS RN 928-51-8

HOCl

CAS RN 928-51-8

IUPAC Name: 4-chlorobutan-1-ol
Synonyms: chlorbutol
Molecular Weight (g/mol): 108.57
Molecular Formula: C4H9ClO
InChi Key: HXHGULXINZUGJX-UHFFFAOYSA-N
SMILES: OCCCCCl
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)