accessibility menu, dialog, popup

UserName

CH3OHC

CAS RN 927-80-0

IUPAC Name: ethoxyethyne
Synonyms: ethoxy-ethyne
Molecular Weight (g/mol): 70.09
Molecular Formula: C4H6O
InChi Key: WMYNMYVRWWCRPS-UHFFFAOYSA-N
SMILES: CCOC#C
Boiling Point 
  • (3)
  • (4)
Molecular Weight (g/mol) 
  • (4)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (3)
  • (4)
  • (4)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (4)