accessibility menu, dialog, popup

UserName

ClOBr

CAS RN 927-58-2

IUPAC Name: 4-bromobutanoyl chloride
Synonyms: 4-bromobutanoyl chloride
Molecular Weight (g/mol): 185.45
Molecular Formula: C4H6BrClO
InChi Key: LRTRXDSAJLSRTG-UHFFFAOYSA-N
SMILES: ClC(=O)CCCBr
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)