CAS RN 92614-55-6
CAS RN 92614-55-6
IUPAC Name:
(2-methylphenyl)methanesulfonyl chloride
Synonyms:
(2-methylphenyl)methanesulfonyl chloride
Molecular Weight (g/mol):
204.67
Molecular Formula:
C8H9ClO2S
InChi Key:
NDMREUGSHNSYBI-UHFFFAOYSA-N
SMILES:
CC1=CC=CC=C1CS(Cl)(=O)=O
1
–
1
of
1
results
1
(2-Methylphenyl)methanesulfonyl chloride, 97%, Thermo Scientific™
CAS: 92614-55-6 Molecular Formula: C8H9ClO2S Molecular Weight (g/mol): 204.668 MDL Number: MFCD02683108 InChI Key: NDMREUGSHNSYBI-UHFFFAOYSA-N Synonym: 2-methylphenyl methanesulfonyl chloride,o-tolylmethanesulfonyl chloride,o-tolyl-methanesulfonyl chloride,o-tolyl methanesulfonyl chloride,o-tolylmethanesulfonylchloride,2-methylbenzylsulphonyl chloride,2-methylphenyl methanesulphonyl chloride,2-methylphenyl methyl sulfonyl chloride,benzenemethanesulfonyl chloride, 2-methyl,chloro 2-methylphenyl methyl sulfone PubChem CID: 2760661 IUPAC Name: (2-methylphenyl)methanesulfonyl chloride SMILES: CC1=CC=CC=C1CS(=O)(=O)Cl