accessibility menu, dialog, popup

UserName

CAS RN 92545-83-0

NOBr

CAS RN 92545-83-0

IUPAC Name: 2-(3-bromophenoxy)pyridine
Synonyms: 2-(3-bromophenoxy)pyridine
Molecular Weight (g/mol): 250.1
Molecular Formula: C11H8BrNO
InChi Key: CXEIVVAAUANDIA-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(OC2=CC=CC=N2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results