accessibility menu, dialog, popup

UserName

H3CMgBr

CAS RN 925-90-6

IUPAC Name: ethylmagnesium bromide
Synonyms: ethylmagnesium bromide
Molecular Weight (g/mol): 133.27
Molecular Formula: C2H5BrMg
InChi Key: FRIJBUGBVQZNTB-UHFFFAOYSA-M
SMILES: [Mg++].[Br-].C[CH2-]
Quantity 
  • (2)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results