CAS RN 923121-46-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-{[5-(Trifluoromethyl)pyridin-2-yl]amino}butanoic Acid, TRC
CAS: 923121-46-4 Molecular Formula: C10H11N2O2F3 Molecular Weight (g/mol): 248.2 IUPAC Name: 4-[[5-(trifluoromethyl)pyridin-2-yl]amino]butanoic acid SMILES: OC(=O)CCCNc1ccc(cn1)C(F)(F)F