accessibility menu, dialog, popup

UserName

CAS RN 922-63-4

CH3H2CO

CAS RN 922-63-4

IUPAC Name: 2-methylidenebutanal
Synonyms: 2-ethylpropenal
Molecular Weight (g/mol): 84.12
Molecular Formula: C5H8O
InChi Key: GMLDCZYTIPCVMO-UHFFFAOYSA-N
SMILES: CCC(=C)C=O
Boiling Point 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results