accessibility menu, dialog, popup

UserName

CAS RN 921939-13-1

SNNHO

CAS RN 921939-13-1

IUPAC Name: [2-(thiophen-2-yl)pyrimidin-5-yl]methanol
Synonyms: [2-(thiophen-2-yl)pyrimidin-5-yl]methanol
Molecular Weight (g/mol): 192.24
Molecular Formula: C9H8N2OS
InChi Key: VPBKFPNQGGDRPA-UHFFFAOYSA-N
SMILES: OCC1=CN=C(N=C1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results