accessibility menu, dialog, popup

UserName

CAS RN 921938-87-6

CH3SOOOOS

CAS RN 921938-87-6

IUPAC Name: [4-(thiophen-2-yl)oxan-4-yl]methyl 4-methylbenzene-1-sulfonate
Synonyms: [4-(thiophen-2-yl)oxan-4-yl]methyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 352.46
Molecular Formula: C17H20O4S2
InChi Key: OZWQPINOYPXHPX-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)S(=O)(=O)OCC1(CCOCC1)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results