accessibility menu, dialog, popup

UserName

CAS RN 921938-78-5

CH3NNOHOH2OHCl

CAS RN 921938-78-5

IUPAC Name: 4-(2-methyl-1H-imidazol-1-yl)benzoic acid hydrate hydrochloride
Synonyms: 4-(2-methylimidazol-1-yl)benzoic acid hydrate hydrochloride
Molecular Weight (g/mol): 256.69
Molecular Formula: C11H13ClN2O3
InChi Key: PSHJNJCAKINUEP-UHFFFAOYSA-N
SMILES: O.Cl.CC1=NC=CN1C1=CC=C(C=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results