accessibility menu, dialog, popup

UserName

H3CCH3

CAS RN 92112-69-1

IUPAC Name: hexane
Synonyms: hexane n-hexane, 2-(13)c-labeled cpd n-hexane, 2-(13)C-labeled cpd 2-(13)C-labeled cpd n-hexane 3-(13)c-labeled cpd n-hexane esani heksan dipropyl 3-(13)C-labeled cpd n-hexane n-hexane, 1-(13)C-labeled cpd
Molecular Weight (g/mol): 86.18
Molecular Formula: C6H14
InChi Key: VLKZOEOYAKHREP-UHFFFAOYSA-N
SMILES: CCCCCC
Quantity 
  • (18)
  • (3)
  • (1)
  • (1)
  • (19)
  • (1)
  • (4)
  • (1)
  • (7)
  • (3)
  • (5)
  • (8)
Grade 
  • (14)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (5)
Percent Purity 
  • (6)
  • (8)
  • (2)
  • (2)
  • (9)
Boiling Point 
  • (3)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (23)
  • (1)
  • (4)
  • (26)
  • (17)
  • (18)
  • (3)
  • (1)
  • (1)
  • (19)
  • (1)
  • (4)
  • (1)
  • (7)
  • (3)
  • (5)
  • (8)
  • (14)
  • (3)
  • (2)
  • (2)
  • (3)
  • (1)
  • (5)
  • (3)
  • (3)
  • (2)
  • (2)
  • (5)
  • (6)
  • (8)
  • (2)
  • (2)
  • (9)
  • (3)
  • (2)
  • (1)
  • (2)