accessibility menu, dialog, popup

UserName

ClCH2N

CAS RN 920-37-6

IUPAC Name: 2-chloroprop-2-enenitrile
Synonyms: 2-propenenitrile, 2-chloro-
Molecular Weight (g/mol): 87.51
Molecular Formula: C3H2ClN
InChi Key: OYUNTGBISCIYPW-UHFFFAOYSA-N
SMILES: ClC(=C)C#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results