accessibility menu, dialog, popup

UserName

NH2

CAS RN 92-67-1

IUPAC Name: [1,1'-biphenyl]-4-amine
Synonyms: 4-aminobiphenyl 4-biphenylnitrenium ion 4-bifenylamin biphenylamine p-xenylamine xenylamine paraaminodiphenyl xenylamin 4-aminodiphenyl ccris 26
Molecular Weight (g/mol): 169.23
Molecular Formula: C12H11N
InChi Key: DMVOXQPQNTYEKQ-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results