accessibility menu, dialog, popup

UserName

H3CN

CAS RN 92-59-1

IUPAC Name: N-benzyl-N-ethylaniline
Synonyms: N-benzyl-N-ethylaniline
Molecular Weight (g/mol): 211.31
Molecular Formula: C15H17N
InChi Key: HSZCJVZRHXPCIA-UHFFFAOYSA-N
SMILES: CCN(CC1=CC=CC=C1)C1=CC=CC=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results