accessibility menu, dialog, popup

UserName

CAS RN 92-55-7

IUPAC Name: (acetyloxy)(5-nitrofuran-2-yl)methyl acetate
Synonyms: (acetyloxy)(5-nitrofuran-2-yl)methyl acetate
Molecular Weight (g/mol): 243.17
Molecular Formula: C9H9NO7
InChi Key: HSXKWKJCZNRMJO-UHFFFAOYSA-N
SMILES: CC(=O)OC(OC(C)=O)C1=CC=C(O1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (9)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (9)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (3)
  • (3)