CAS RN 92-45-5
IUPAC Name:
3-chloro-2H-chromen-2-one
Synonyms:
3-chlorochromen-2-one
Molecular Weight (g/mol):
180.59
Molecular Formula:
C9H5ClO2
InChi Key:
CKCOPMSBJBNBCQ-UHFFFAOYSA-N
SMILES:
ClC1=CC2=CC=CC=C2OC1=O
1
–
1
of
1
results
1
3-Chlorocoumarin, 97%, Thermo Scientific™
CAS: 92-45-5 Molecular Formula: C9H5ClO2 Molecular Weight (g/mol): 180.59 MDL Number: MFCD00024073 InChI Key: CKCOPMSBJBNBCQ-UHFFFAOYSA-N PubChem CID: 96133 IUPAC Name: 3-chlorochromen-2-one SMILES: ClC1=CC2=CC=CC=C2OC1=O