accessibility menu, dialog, popup

UserName

CAS RN 92-24-0

CAS RN 92-24-0

IUPAC Name: tetracene
Synonyms: tetracene 2,3-benzanthrene chrysogen 2,3-benzanthracene tetracene (hydrocarbon) benz(b)anthracene rubene
Molecular Weight (g/mol): 228.29
Molecular Formula: C18H12
InChi Key: IFLREYGFSNHWGE-UHFFFAOYSA-N
SMILES: C1=CC2=CC3=CC4=CC=CC=C4C=C3C=C2C=C1
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)