CAS RN 91987-74-5
CAS RN 91987-74-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
DOTP, TRC
CAS: 91987-74-5 Molecular Formula: C12H32N4O12P4 Molecular Weight (g/mol): 548.3 Synonym: 1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetra(methylene Phosphonic Acid),1,4,7,10-Tetraazacyclododecane, Phosphonic Acid deriv.,1,4,7,10-Tetraazacyclododecane-1,4,7,10-tetramethylenephosphonic Acid,DOTMP,NSC 681106,N,N',N'',N'''-Tetrakis(phosphonomethyl)-1,4,7,10-tetraazacyclododecane SMILES: OP(=O)(O)CN1CCN(CP(=O)(O)O)CCN(CP(=O)(O)O)CCN(CP(=O)(O)O)CC1