CAS RN 918-01-4
CAS RN 918-01-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bromodichloronitromethane, TRC
CAS: 918-01-4 Molecular Formula: C Br Cl2 N O2 Molecular Weight (g/mol): 208.83 Synonym: Bromodichloronitromethane (ACI),Dichlorobromonitromethane IUPAC Name: bromo-dichloro-nitromethane SMILES: [O-][N+](=O)C(Cl)(Cl)Br