accessibility menu, dialog, popup

UserName

KH3CO

CAS RN 917-58-8

IUPAC Name: potassium ethanolate
Synonyms: potassium ethoxide
Molecular Weight (g/mol): 84.16
Molecular Formula: C2H5KO
InChi Key: RPDAUEIUDPHABB-UHFFFAOYSA-N
SMILES: [K+].CC[O-]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)