accessibility menu, dialog, popup

UserName

CAS RN 916766-84-2

CH3NONOH

CAS RN 916766-84-2

IUPAC Name: [3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol
Synonyms: [3-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methanol
Molecular Weight (g/mol): 190.2
Molecular Formula: C10H10N2O2
InChi Key: ZBIYTANHMLGZFB-UHFFFAOYSA-N
SMILES: CC1=NOC(=N1)C1=CC(CO)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results