accessibility menu, dialog, popup

UserName

H3COFFFN

CAS RN 916421-02-8

IUPAC Name: 2-[3-methoxy-5-(trifluoromethyl)phenyl]acetonitrile
Synonyms: 2-[3-methoxy-5-(trifluoromethyl)phenyl]acetonitrile
Molecular Weight (g/mol): 215.18
Molecular Formula: C10H8F3NO
InChi Key: IZJPZDDRQKCSEL-UHFFFAOYSA-N
SMILES: COC1=CC(CC#N)=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results