accessibility menu, dialog, popup

UserName

CAS RN 916420-59-2

CH3NOFFF

CAS RN 916420-59-2

IUPAC Name: 3-methyl-5-(trifluoromethoxy)benzonitrile
Synonyms: 3-methyl-5-(trifluoromethoxy)benzonitrile
Molecular Weight (g/mol): 201.15
Molecular Formula: C9H6F3NO
InChi Key: RUVNGFLRCPBNJK-UHFFFAOYSA-N
SMILES: CC1=CC(OC(F)(F)F)=CC(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results