accessibility menu, dialog, popup

UserName

CH3ONH2OFFF

CAS RN 916420-53-6

IUPAC Name: 3-methyl-5-(trifluoromethoxy)benzamide
Synonyms: 3-methyl-5-(trifluoromethoxy)benzamide
Molecular Weight (g/mol): 219.16
Molecular Formula: C9H8F3NO2
InChi Key: QKDMEDIWAQNUHI-UHFFFAOYSA-N
SMILES: CC1=CC(OC(F)(F)F)=CC(=C1)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results