CAS RN 916210-98-5
CAS RN 916210-98-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(2-Chloropyridin-4-yl)methanamine hydrochloride, MedChemExpress
MedChemExpress (2-Chloropyridin-4-yl)methanamine hydrochloride is a selective LOXL2 inhibitor with an IC50 of 126 nM.