accessibility menu, dialog, popup

UserName

CAS RN 916210-69-0

NH2OFFFCl

CAS RN 916210-69-0

IUPAC Name: 1-[4-chloro-3-(trifluoromethoxy)phenyl]methanamine
Synonyms: 1-[4-chloro-3-(trifluoromethoxy)phenyl]methanamine
Molecular Weight (g/mol): 225.6
Molecular Formula: C8H7ClF3NO
InChi Key: XZZFTTABTJCTSR-UHFFFAOYSA-N
SMILES: NCC1=CC(OC(F)(F)F)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results