accessibility menu, dialog, popup

UserName

CAS RN 91600-33-8

IUPAC Name: ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutanoate
Synonyms: ethyl 4,4,4-trifluoro-3-hydroxy-2-methylbutanoate
Molecular Weight (g/mol): 200.16
Molecular Formula: C7H11F3O3
InChi Key: MFRIOKNLYRUYHP-UHFFFAOYNA-N
SMILES: CCOC(=O)C(C)C(O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results