accessibility menu, dialog, popup

UserName

CAS RN 915707-56-1

H3CNNNH2

CAS RN 915707-56-1

IUPAC Name: 1-[2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanamine
Synonyms: 1-[2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanamine
Molecular Weight (g/mol): 219.33
Molecular Formula: C13H21N3
InChi Key: YLJBTEMLBKZLIT-UHFFFAOYSA-N
SMILES: CN1CCCN(CC1)C1=CC=CC=C1CN

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results