accessibility menu, dialog, popup

UserName

CAS RN 915707-55-0

H3CNNOH

CAS RN 915707-55-0

IUPAC Name: [2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanol
Synonyms: [2-(4-methyl-1,4-diazepan-1-yl)phenyl]methanol
Molecular Weight (g/mol): 220.32
Molecular Formula: C13H20N2O
InChi Key: IYBDLMFCIKDQDV-UHFFFAOYSA-N
SMILES: CN1CCCN(CC1)C1=CC=CC=C1CO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results