accessibility menu, dialog, popup

UserName

NSNH2Cl

CAS RN 91447-89-1

IUPAC Name: 2-chloropyridine-4-carbothioamide
Synonyms: 2-chloropyridine-4-carbothioamide
Molecular Weight (g/mol): 172.63
Molecular Formula: C6H5ClN2S
InChi Key: SXOBQZFUVLZZCG-UHFFFAOYSA-N
SMILES: NC(=S)C1=CC(Cl)=NC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results