accessibility menu, dialog, popup

UserName

HNNHOO

CAS RN 914348-04-2

IUPAC Name: benzyl N-(azetidin-3-yl)carbamate
Synonyms: benzyl N-(azetidin-3-yl)carbamate
Molecular Weight (g/mol): 206.25
Molecular Formula: C11H14N2O2
InChi Key: UWOOBFRZDNDUQB-UHFFFAOYSA-N
SMILES: O=C(NC1CNC1)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results