accessibility menu, dialog, popup

UserName

CAS RN 91424-40-7

H3CH3CCH3SiH3CCH3OOOO

CAS RN 91424-40-7

IUPAC Name: 4-[(tert-butyldimethylsilyl)oxy]oxane-2,6-dione
Synonyms: 4-[(tert-butyldimethylsilyl)oxy]oxane-2,6-dione
Molecular Weight (g/mol): 244.36
Molecular Formula: C11H20O4Si
InChi Key: RXAJGRHLLRGVSB-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1CC(=O)OC(=O)C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results