accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CClNOO

CAS RN 913836-26-7

IUPAC Name: 2-(4-chloro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(4-chloro-3-nitrophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 283.52
Molecular Formula: C12H15BClNO4
InChi Key: VEIRGLFECZUCTI-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(=C(Cl)C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results