accessibility menu, dialog, popup

UserName

CAS RN 913836-12-1

BOH3COOHHOOH

CAS RN 913836-12-1

IUPAC Name: 5-(dihydroxyboranyl)-2-methoxybenzoic acid
Synonyms: 5-(dihydroxyboranyl)-2-methoxybenzoic acid
Molecular Weight (g/mol): 195.97
Molecular Formula: C8H9BO5
InChi Key: YZKWFWNYFKBAHO-UHFFFAOYSA-N
SMILES: COC1=C(C=C(C=C1)B(O)O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results